missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-10 R alpha Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF-474-NA
This item is not returnable.
View return policy
Description
IL-10R alpha Polyclonal specifically detects IL-10R alpha in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-10R alpha | |
| Polyclonal | |
| Unconjugated | |
| Q61727 | |
| IL10RA | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-10 R alpha Thr24-Asn238 (Lys188Arg and Glu222Gly) Accession # Q61727 | |
| 100 μg | |
| Primary | |
| Detects mouse IL-10 R alpha in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 25% cross-reactivity with recombinant human IL-10 R alpha is observed. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 1-3 ug/mL | |
| CD210 antigen, CDw210a, HIL-10R, IBD28, IL-10 receptor subunit alpha, IL10R alpha, IL-10R subunit 1, IL-10R1, IL10RA, IL-10RA, IL10RIL-10R subunit alpha, interleukin 10 receptor, alpha, interleukin-10 receptor alpha chain, Interleukin-10 receptor subunit 1, interleukin-10 receptor subunit alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3587 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction