missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-10 (ELISA Standard) Recombinant Protein

Product Code. 15508726
Click to view available options
Quantity:
4 pack
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 15508726

Brand: Invitrogen™ 29810165

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Description: mouse IL-10 is an ~18 kDa cytokine, also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response. Applications Reported: The Mouse IL-10 Lyophilized Standard is intended to be used as a standard for the Mouse IL-10 Ready-Set-Go! ELISA kit (Product # 88-7105). Typically the standard is run in a series of 7 doubling dilutions from the 'top' standard. Please refer to the Certificate of Analysis included with each ELISA set for instructions on the proper reconstitution and dilution of the lyophilized standard. Note: These are single-use vials. Use one time within 4 hours of reconstitution and discard. Reconstitute and dilute as suggested in the Certificate of Analysis or recovery may not be accurate. Applications Tested: The Mouse IL-10 Lyophilized Standard has been calibrated and tested in the Mouse IL-10 Ready-Set-Go! ELISA kit (Product # 88-7105-76). Source: E. coli expressed amino acids Ser 19-Ser 178 of mature mouse IL-10 accession # NM_010548 with a Cys149tyr substitution.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18893
For Use With (Application) Neutralization, ELISA Standard
Formulation Protein with no preservative
Gene ID (Entrez) 16153
Molecular Weight (g/mol) 18 kDa
Name Mouse IL-10 (ELISA Standard)
Purification Method Purified
Quantity 4 pack
Source E. Coli
Storage Requirements 4°C
Regulatory Status RUO
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Common Name IL-10
Gene Symbol IL10
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Mouse IL-10, amino acids Ser19-Ser178 (Accession # NM_010548) with a Cys149Tyr substitution
Content And Storage 4°C
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.