missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-10 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 36 publications
Brand: R&D Systems MAB417-500
This item is not returnable.
View return policy
Description
IL-10 Monoclonal specifically detects IL-10 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3586 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| JES052A5 | |
| Western Blot 1 ug/mL, Neutralization 0.005-0.015 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P18893 | |
| IL10 | |
| E. coli-derived recombinant mouse IL-10 Accession # P18893 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-10 in sandwich ELISAs and Western blots. In sandwich ELISAs, less than 8% cross-reactivity with recombinant rat IL-10 and 1% cross-reactivity with recombinant cotton rat IL-10 is observed. In sandwich ELISAs, no cross-reactivity with recombinant human (rh) IL-10, recombinant mouse IL-10 R, rhIL-10 R, recombinant canine IL-10, recombinant feline IL-10, or recombinant porcine IL-10 is observed. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction