missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1 RII Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF563
This item is not returnable.
View return policy
Description
IL-1 RII Polyclonal specifically detects IL-1 RII in Mouse samples. It is validated for Western Blot.
Specifications
| IL-1 RII | |
| Polyclonal | |
| Biotin | |
| P27931 | |
| IL1R2 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-1 RII Met1-Glu355 Accession # P27931 | |
| 50 μg | |
| Primary | |
| Detects mouse IL-1 RII in Western blots. In Western blots, less than 1% cross-reactivity with human IL-1 RII, mouse IL-18 R, human IL-1 RAcp, rat IL-1 Rrp2, and mouse IL-1 RI is observed and no cross-reactivity with mouse IL-18 AcpLR is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| beta, CD121b, CD121b antigen, IL-1 R beta, IL1R2, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL1RII, IL-1RII, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7850 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction