missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1 RI PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£242.00 - £442.00
Specifications
| Antigen | IL-1 RI |
|---|---|
| Clone | 129304 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721829
![]() |
R&D Systems
FAB7712P-025 |
25 Tests |
£242.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18726756
![]() |
R&D Systems
FAB7712P-100 |
100 Tests |
£442.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Mouse samples. It is validated for Flow Cytometry.Specifications
| IL-1 RI | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| IL1R1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse IL-1 RI in direct ELISAs and Western blots. |
| 129304 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Mouse | |
| P13504 | |
| 3554 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI Leu20-Lys338 Accession # P13504 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title