missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1 RI Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB7712G-100UG
This item is not returnable.
View return policy
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-1 RI | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P13504 | |
| IL1R1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI Leu20-Lys338 Accession # P13504 | |
| RUO | |
| 3554 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 129304 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse IL-1 R1 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction