missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN gamma, Recombinant Protein, Carrier-Free, Invitrogen™
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Human IFN gamma is an ∽20 kDa factor produced by activated T, B and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN-γ. Glycosylation of IFN gamma does not affect its biological activity.
Specifications
Specifications
| Accession Number | NM_000619 |
| Concentration | 0.5mg/mL |
| For Use With (Application) | Cytokine Bioassay |
| Formulation | Sterile liquid: 20mM phosphate, 400mM NaCl, pH 7.2, 0.22μm filtered. |
| Molecular Weight (g/mol) | The methionylated N-terminal protein migrates as a 16kDa polypeptide on SDS-PAGE. |
| Name | IFN gamma |
| Purification Method | SDS-PAGE |
| Quantity | 100 μg |
| Source | E. coli |
| Storage Requirements | Store at less than or equal to -70°C. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction