missing translation for 'onlineSavingsMsg'
Learn More

Annexin V, Recombinant Protein, PE, eBioscience™

Product Code. 15551917
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
This item is not returnable. View return policy

Product Code. 15551917

Brand: Invitrogen BMS306PE100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

The protein encoded by this gene belongs to the annexin family of calcium-dependent phospholipid binding proteins some of which have been implicated in membrane-related events along exocytotic and endocytotic pathways. Annexin 5 is a phospholipase A2 and protein kinase C inhibitory protein with calcium channel activity and a potential role in cellular signal transduction, inflammation, growth and differentiation. Annexin 5 has also been described as placental anticoagulant protein I, vascular anticoagulant-alpha, endonexin II, lipocortin V, placental protein 4 and anchorin CII. The gene spans 29 kb containing 13 exons, and encodes a single transcript of approximately 1.6 kb and a protein product with a molecular weight of about 35 kDa. Description: Annexins are a family of calcium-dependent phospholipid-binding proteins, which bind to phosphatidylserine (PS) to identify apoptotic cells. In healthy cells, PS is predominantly located along the cytosolic side of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution in the phospholipid bilayer and translocates to the extracellular membrane, which is detectable with fluorescently labeled Annexin V. In early stages of apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI) and 7-AAD, therefore cells which display only Annexin V staining (PI/7-AAD negative) are in early stages of apoptosis. During late-stage apoptosis, loss of cell membrane integrity allows Annexin V binding to cytosolic PS, as well as cell uptake of PI and 7-AAD. Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry. Annexin V has also been identified as an anticoagulant protein in the blood coagulation cascade, by acting as an inhibitor of prothrombin activation. The presence of antibodies to Annexin V is associated with systemic lupus erythematosus (SLE), recurrent spontaneous abortions and systemic sclerosis (SSc). Purity: Annexin V is >89% pure as demonstrated by SDS-gel electrophoresis and reverse phase HPLC. The fluorochrome phycoerythrin (PE) shows an excitation maximum of 495 to 564 nm and an emission maximum of 576 nm. Annexin V conjugated to PE is suitable for the detection of PS and allows simple secondary labelling by staining, for example, the membrane surface proteins with a monoclonal antibody for further cellular characterization.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08758, P48036, P14668
Concentration 5μL/Test
For Use With (Application) Flow Cytometry
Formulation TBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene ID (Entrez) 308
Molecular Weight (g/mol) 35.8kDa
Name Annexin V
pH Range 7.4
Purification Method SDS-gel electrophoresis and reverse phase HPLC
Quantity 100 Tests
Source E. coli
Storage Requirements 4°C, do not freeze
Regulatory Status RUO
Structural Form E. Coli
Common Name Annexin V
Gene Symbol ANXA5
Biological Activity Annexin V exhibits anti-phospholipase activity and binds to phosphatidylserine (PS).
Product Type Recombinant Protein
Specific Reactivity All
Conjugate PE Labeled
Cross Reactivity All
Species E. coli
Recombinant Recombinant
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade Affinity chromatography
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.