missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-1 alpha ELISA Kits

Product Code. 10209193
Change view
Click to view available options
Quantity:
1 Plate Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10209193 1 Plate Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10209193 Supplier Invitrogen™ Supplier No. EMIL1A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Measure levels of mouse interleukin-1 alpha (IL-1a) in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates.

Assay IL-1aα (mouse interleukin-1 alpha) levels in mouse serum, plasma (EDTA, heparin, sodium citrate)and culture supernatant samples with our Thermo Scientific Mouse IL-1 alpha ELISA Kits.

These mouse IL-1 alpha ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-1 alpha (IL-1α) in samples at 50 per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant mouse interleukin-1 alpha (IL-1α)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 15.6 to 1000pg/mL

  • Sensitivity: < 6pg/mL

  • Precision: Intra-assay CV ≤ 10%; Inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, human IL-1β, or mouse IL-1β, IL-3, IL-4, IL-5, IL-7, IL-10, TNFα, IFNγ, or GM-CSF

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.6 to 1000pg/mL
Assay Sensitivity <6pg/mL
Description Mouse IL-1 alpha ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity No significant cross-reaction with human IL-1a, human IL-1ß, or mouse IL-1ß, IL-3, IL-4, IL-5, IL-7, IL-10, TNFa, IFNg, or GM-CSF
Sufficient For 96 microplate wells
Kit Contents Biotinylated Antibody Reagent, 8mL, Anti-Mouse IL-1 alpha Precoated 96-well Strip Plate, 1 ea, Adhesive Plate Covers, 6 ea, 30X Wash Buffer, 50mL, HRP Dilution Buffer, 14mL, Streptavidin-HRP Concentrate, 75 μL, Lyophilized Recombinant Mouse IL-1 alpha Standard, 2 vials, TMB Substrate Solution, 13mL, Stop Solution, 13mL, Standard Diluent, 12mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV ≤ 10%; Inter-assay CV ≤ 10%
Quantity 1 Plate Kit
Sample Volume 50 μL
Target Natural and recombinant mouse interleukin-1 alpha (IL-1a)
Total Assay Time 3 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.