missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IGSF6/DORA (NP_109616) VersaClone cDNA
Click to view available options
:
10 μg
Description
- IGSF6 is a member of the immunoglobulin-like domaincontaining superfamily.
- Proteins in this superfamily contain varying numbers of immunoglobulin-like domains and are thought to participate in the regulation of interactions between cells.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mIgsf6 |
| Accession Number | NP_109616 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 727 bp |
| Gene ID (Entrez) | 10261 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction