missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IGSF2/CD101 Antibody, R&D Systems™
Rat Monoclonal Antibody
£363.92
Specifications
Antigen | IGSF2/CD101 |
---|---|
Clone | 307707 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
IGSF2/CD101 Monoclonal specifically detects IGSF2/CD101 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
IGSF2/CD101 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Rat | |
Mouse | |
A8E0Y8 | |
9398 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse IGSF2/CD101 Gln21-Tyr1033 Accession # A8E0Y8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse IGSF2/CD101. |
307707 | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD101, CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, Leukocyte Surface Protein, member 2, V7 | |
CD101 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only