missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgM Isotype Control, PE, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen™ MGM04
Code : 50
Additional Details : Weight : 0.50000kg
Description
Mouse IgM Isotype Control for Flow
Mouse IgM Control antibody specifically detects Mouse IgM in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgM | |
PE | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgM | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests |