Learn More
Mouse IgG3 PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC007P
This item is not returnable.
View return policy
Description
IgG3 Monoclonal specifically detects IgG3 in Mouse samples. It is validated for Control.
Specifications
| IgG3 | |
| 133316 | |
| Control 10 uL/10^6 cells | |
| DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| 3502 | |
| Mouse | |
| IgG3 |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IGHG3 | |
| KLH | |
| 200 Tests | |
| Selected for use as mouse IgG 3 isotype control for flow cytometry applications. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |