Learn More
Mouse IgG2B PE-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 33 publications
Brand:  R&D Systems IC0041P
Product Code. 18123961
  • £276.00 / 200 tests
Description
IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG2b
133303
Control 10 uL/10^6 cells
Mouse
Protein A or G purified from hybridoma culture supernatant
Selected for use as mouse IgG 2B isotype control for flow cytometry applications.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
KLH
200 Tests
Mouse
IgG2b