Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), FITC, eBioscience™, Invitrogen™
Mouse Isotype Control
£65.50 - £106.00
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Concentration | 0.5mg/mL |
Applications | Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15288219
|
Affymetrix eBioscience
11-4732-81 |
50 μg |
£106.00
50µg |
In Stock
Log in to see stock. |
|||||
15556536
|
Affymetrix eBioscience
11-4732-41 |
25 Tests |
£65.50
25 tests |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15566536
|
Affymetrix eBioscience
11-4732-42 |
100 Tests |
£106.00
100 tests |
Estimated Shipment: 09-05-2024
Log in to see stock. |
|||||
15298219
|
Affymetrix eBioscience
11-4732-82 |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: This mouse IgG2b isotype control has been reported for use in immunocytochemistry, immunohistochemistry, and flow cytometric analysis. Applications Tested: FITC Mouse IgG2b Isotype Control is offered in 2 formats: - µg size: has been tested by flow cytometric analysis and can be used at the same concentration as the experimental antibody.- test size: has been pre-titrated and tested by intracellular flow cytometric analysis and can be used at test size: 5 µL (1 µg)/test per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Mouse IgG2b is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the third most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2b is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2b is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2b kappa | |
0.5mg/mL | |
FITC | |
Mouse | |
Not Applicable | |
IgG2, IgG2b, IgG2bk | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBMG2b | |
Control, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Liquid | |
RUO | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.