missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b Isotype Control (M2B), Texas Red, Novus Biologicals™

Product Code. 18058756 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18058756 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18058756 Supplier Novus Biologicals Supplier No. NBP196795

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2b
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Conjugate Texas Red
Dilution Flow Cytometry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Host Species Mouse
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Isotype Control (Primary)
Test Specificity Isotyping assay resulted the material is non-reactive with antisera to mouse IgG1, IgG2a, IgG3, IgM, and IgA. Texas Red(TM) Conjugated Mouse IgG2b was proven by Dot Blot.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent).
Target Species Mouse
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.