missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG2a Isotype Control (PPV-04), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA110418
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody will not cross-react with human, porcine, mouse or rat.
Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG2a | |
PPV-04 | |
Unconjugated | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Plum pox virus | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG2a |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |