missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgG1 Mouse ELISA Standard
Brand: Invitrogen™ 39-50410-65
Code : NEW
Additional Details : Weight : 0.47500kg
Description
Component of Mouse IgG1 ELISA Ready-SET-Go! KitSpecifications
Kit | |
Recombinant | |
eBioscience | |
Mouse | |
Mouse | |
Liquid |
4 Pack | |
Microplate Reader | |
ELISA Standard | |
IgG1 | |
Mouse |
For Research Use Only. Not for use in diagnostic procedures.