missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PerCP-Cyanine5.5, eBioscience™, Invitrogen™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 45-4714-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15341180

  • £72.75 / 25µg
In Stock
to see stock.
Add to basket

Description

Description

Description: The monoclonal mouse IgG1 K immunoglobulin is useful as an isotype control. Applications Reported: The PerCP-Cy5.5 Mouse IgG1 K Isotype Control has been reported for use in flow cytometric analysis. Applications Tested: The Mouse IgG1 K Isotype Control has been tested by flow cytometric analysis. Use isotype control at the same concentration as experimental antibody. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
Specifications

Specifications

Mouse IgG1 kappa
P3.6.2.8.1
PerCP-Cyanine5.5
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
25 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.