Learn More
Mouse IgG1 Isotype Control, FITC, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA518096
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-18096 is negative on all human cells and cell lines tested. This antibody recognizes a rat cell surface marker, and therefore cannot be used as a negative control in this species. Test results have shown that MA5-18096 is also suitable for use as a negative control with bovine, ovine, porcine, equine, canine, lapine and guinea-pig tissues. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG1 | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
FITC | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Rat thymocyte membrane glycoproteins | |
100 Tests | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG1 |