missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Assay Range | 0.16 to 10 ug/mL |
| Assay Sensitivity | 0.09 ug/mL |
| Product Type | ELISA Kit (Chemiluminescence) |
| Sample Type | Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Rat IgG1 in samples. No significant cross-reactivity or interference between Rat IgG1 and analogues was observed. |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | IgG1, Igh-4, RGD1359539, VH7183 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?