Learn More
Mouse IgG1 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems IC002A
Product Code. 18114781
  • £276.00 / 200 tests
Description
IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG1
11711
Control 10 uL/10^6 cells
IgG1, Igh-4, RGD1359539, VH7183
Mouse
Protein A or G purified from hybridoma culture supernatant
Selected for use as mouse IgG 1 isotype control for flow cytometry applications.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
IGHG1
KLH
200 Tests
Mouse
IgG1 κ