missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE

Goat Polyclonal Antibody

£271.00 - £2155.00

Specifications

Antigen IgG
Dilution Immunohistochemistry
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18772848
GREENER_CHOICE
View Documents
R&D Systems
VC001-025
25 mL
£271.00
25mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18740628
GREENER_CHOICE
View Documents
R&D Systems
VC001-050
50 mL (2 x 25 mL) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18764388
GREENER_CHOICE
Ver documentos
R&D Systems
VC001-125
125 mL
£1140.00
125mL
Ver productos alternativos
Ahorre hasta 
»
Please to purchase this item. Need a web account? Register with us today!
18791488
GREENER_CHOICE
Ver documentos
R&D Systems
VC001-250
250 mL (2 x 125 mL)
£2155.00
250mL
Ver productos alternativos
Ahorre hasta 
»
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Immunohistochemistry.
Especificaciones

Especificaciones

IgG
Immunohistochemistry
HRP Polymer
RUO
Supplied as a solution in PBS with preservative.
3500
IgG
Affinity Purified
Detects heavy ( gamma ) chains on mouse IgG and light chains on all mouse immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin mouse serum proteins or serum proteins from bovine, horse, human, pig or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
Immunohistochemistry
Polyclonal
Goat
Mouse
Immunoglobulin G, ImmunoglobulinG
Mouse IgG
Secondary
6 months from date of receipt, 2 to 8 degreesC as supplied.
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado