missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgA Mouse ELISA Standard
Click to view available options
Quantity:
4 pack
Unit Size:
Pack of 4
Description
Component of Mouse IgA ELISA Ready-SET-Go! Kit
Specifications
Specifications
| Format | Kit |
| Quantity | 4 pack |
| Recombinant | Recombinant |
| For Use With (Equipment) | Microplate Reader |
| Former Identifiers | eBioscience |
| Immunoassay Kit Format | ELISA Standard |
| Target Species Primary | Mouse |
| Target Kit Named | IgA |
| Target Species Named | Mouse |
| Target Species Validated | Mouse |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction