missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-alpha/beta R1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3039-SP
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Polyclonal specifically detects IFN-alpha/beta R1 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| IFN-alpha/beta R1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFN-aR1, IFNAR1, IFNBR, IFNbR1, IFN-bR1, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3454 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 1-4 ug/mL | |
| P33896 | |
| IFNAR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IFN-alpha/beta R1 Glu27-Thr429 Accession # P33896 | |
| 25 μg | |
| Primary | |
| Detects mouse IFN-alpha/beta R1 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant mouse (rm) IFN-alpha/beta R2, rmIFN-gamma R1, and rmIFN-gamma R2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction