missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IFN-alpha 2 Recombinant Protein, eBioscience™

Product Code. 15567516
Click to view available options
Quantity:
25 μg
5 μg
Unit Size:
25µg
5µg
This item is not returnable. View return policy

Product Code. 15567516

Brand: Invitrogen™ 14831262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01573
Concentration 0.1 mg/mL
For Use With (Application) Bioactivity, Functional Assay
Formulation 50mM sodium acetate with 0.1M NaCl, 1% BSA and no preservative; pH 5.0
Gene ID (Entrez) 15965
Molecular Weight (g/mol) 19.4 kDa
Name Mouse IFN-alpha 2
pH Range 5.0
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -80°C
Regulatory Status RUO
Endotoxin Concentration <0.01 ng/μg
Gene Alias alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2
Common Name Interferon alpha-2
Gene Symbol IFNA2
Biological Activity ED50 = 0.1 ng/mL; as measured by the inhibition of the cytopathic effect of EMC virus on L929 cells.
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence Mature mouse IFN-alpha 2, amino acids Cys24-Glu190 (Accession # NM_010503)
Content And Storage -80°C
Activity ED50 = 0.1 ng/mL; as measured by the inhibition of the cytopathic effect of EMC virus on L929 cells.
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >98% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.