missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ICOS Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB168G-100UG
This item is not returnable.
View return policy
Description
ICOS Monoclonal specifically detects ICOS in Mouse samples. It is validated for Flow Cytometry.
Specifications
| ICOS | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9WVS0 | |
| ICOS | |
| NS0 mouse myeloma cell line transfected with mouse ICOS Accession # Q9WVS0 | |
| RUO | |
| 29851 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 670306 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse ICOS in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction