missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ICAM-1/CD54 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 43 publications
Brand: R&D Systems AF796-SP
This item is not returnable.
View return policy
Description
ICAM-1/CD54 Polyclonal specifically detects ICAM-1/CD54 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Adhesion Blockade.
Specifications
| ICAM-1/CD54 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM1, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3383 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Adhesion Blockade | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Adhesion Blockade | |
| Q3U8M7 | |
| ICAM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ICAM-1/CD54 Gln28-Asn485 Accession # Q3U8M7 | |
| 25 μg | |
| Primary | |
| Detects mouse ICAM-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant rat ICAM-1 is observed, approximately 5% cross-reactivity with recombinant human (rh) ICAM-1 and rhICAM-3 is observed, and less than 1% cross-reactivity with recombinant mouse ICAM-2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction