missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HGFR/c-MET Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB5271
This item is not returnable.
View return policy
Description
HGFR/c-MET Monoclonal specifically detects HGFR/c-MET in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| HGFR/c-MET | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4233 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 118627 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P16056 | |
| MET | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse HGF R Glu25-Asn929 Accession # P16056 | |
| 500 μg | |
| Primary | |
| Detects mouse HGF R in ELISAs. In a sandwich ELISA, no cross-reactivity or interference was observed with recombinant human (rh) HGF R, rmHGFA, or rhMSP R. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction