missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5406-SP
This item is not returnable.
View return policy
Description
Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 Polyclonal specifically detects Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Heparan Sulfate 6-O-Sulfotransferase 3/HS6ST3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp761K2315, heparan sulfate 6-O-sulfotransferase 3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 266722 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| NP_056635 | |
| HS6ST3 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Heparan Sulfate 6-O-Sulfotransferase/HS6ST3 Pro28-Trp470 Accession # NP_056635 | |
| 25 μg | |
| Primary | |
| Detects mouse Heparan Sulfate 6-O-Sulfotransferase/HS6ST3 in direct ELISAs. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction