missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Hemopexin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7007
This item is not returnable.
View return policy
Description
Hemopexin Monoclonal specifically detects Hemopexin in Mouse samples. It is validated for Western Blot.
Specifications
| Hemopexin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q91X72 | |
| HPX | |
| Mouse myeloma cell line NS0-derived recombinant mouse Hemopexin Met1-Gln460 Accession # Q91X72 | |
| 100 μg | |
| Primary | |
| Detects mouse Hemopexin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Hemopexin is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 781941 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-1B-glycoprotein, FLJ56652, hemopexin, HPX, HX | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3263 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction