Learn More
Invitrogen™ Mouse Hematopoietic Lineage Antibody Cocktail, FITC, eBioscience™
Antibody Cocktail
Brand: Invitrogen™ 22-7770-72
Description
Description: The Mouse Hematopoietic Lineage FITC Cocktails contains FITC-conjugated antibodies that can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on the expression of cell surface antigens. This cocktail contains the following antibodies: Anti-Mouse CD3 (17A2) FITC Anti-Mouse CD45R (B220) (RA3-6B2) FITC Anti-Mouse CD11b (M1/70) FITC Anti-Mouse TER-119 (TER-119) FITC Anti-Mouse Ly-G6 (Gr-1) (RB6-8C5) FITC Applications Reported: The Mouse Hematopoietic Lineage FITC Cocktail has been reported for use in flow cytometric analysis. Applications Tested: The Mouse Hematopoietic Lineage FITC Cocktail has been pre-titrated and tested by flow cytometric analysis of C57Bl/6 bone marrow cells. This can be used at 20 μL per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Hematopoietic Lineage antibodies can be used to identify, enrich and/or deplete cells committed to the T, B, NK, myeloid and erythroid lineages based on their expression of cell surface antigens.
Specifications
| Mouse Hematopoietic Lineage | |
| Cocktail | |
| 20 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Rat | |
| 50 Tests | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 17A2, RA3-6B2, M1/70, TER-119, RB6-8C5, RA3-6B2, M1/70, TER-119, RB6-8C5 | |
| FITC | |
| 17A2; Hematopoietic Lineage Cocktail; M1/70; RA3-6B2; RB6-8C5; TER-119 | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Antibody | |
| Responsibly packaged |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.