missing translation for 'onlineSavingsMsg'
Learn More

Abcam Mouse heart tissue lysate - mitochondrial extract, 2 mg

Product Code. 32150985 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
32150985 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32150985 Supplier Abcam Supplier No. ab1103502mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse heart cell/tissue lysate.

  • Species: Mouse
  • Tissue: heart
  • Form: Liquid

Specifications

For Use With (Application) Western Blot, SDS-PAGE, Immunoprecipitation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.