missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HAI-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1107-SP
This item is not returnable.
View return policy
Description
HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for Western Blot.
Specifications
| HAI-2/SPINT2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10653 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 170317 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SPINT2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse HAI-2B aa 30-197 (del 37-93) | |
| 25 μg | |
| Primary | |
| Detects rmHAI-2 in direct ELISAs and Western blots. Less than 5% cross-reactivity was observed with rmHAI-1 and rhHAI-2. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction