missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse HAI-2 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1107
Additional Details : Weight : 0.00970kg
Description
HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for Western Blot.Specifications
HAI-2/SPINT2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
HAI2, HAI2A, HAI2B, HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2, SPINT2 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10653 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
170317 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
SPINT2 | |
Mouse myeloma cell line NS0-derived recombinant mouse HAI-2B aa 30-197 (del 37-93) | |
500 μg | |
Primary | |
Detects rmHAI-2 in direct ELISAs and Western blots. Less than 5% cross-reactivity was observed with rmHAI-1 and rhHAI-2. | |
Mouse | |
Purified |
For Research Use Only