Learn More
Gibco™ Mouse Granzyme B Recombinant Protein, PeproTech®
Description
140-03-1MG will be provided as 10 x 100 μg (140-03-100UG). 140-03-250UG will be provided as multiple 50 μg (140-03-50UG) and 100 μg (140-03-100UG) vials. 140-03-500UG will be provided as 5 x 100 μg (140-03-100UG). Recombinant Murine Granzyme B is a glycosylated 227 amino acid protein, comprising the mature active portion of the murine Granzyme B precursor. The apparent molecular weight is 28.9 kDa by mass spectrometry. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P04187 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 14939 |
| Molecular Weight (g/mol) | 28.9 |
| Name | Mouse Granzyme B |
| Quantity | 5 x 100 μg |
| Source | Baculovirus |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.