missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Gr-1/Ly-6G PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
£204.00 - £396.00
Specifications
| Antigen | Gr-1/Ly-6G |
|---|---|
| Clone | RB6-8C5 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
18746369
![]() |
R&D Systems
FAB1037P-025 |
25 Tests |
£204.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18767786
![]() |
R&D Systems
FAB1037P-100 |
100 Tests |
£396.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.Spezifikation
| Gr-1/Ly-6G | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 58530 | |
| The immunogen for this antibody is Ly-6G (Gr-1, Gr1). | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| RB6-8C5 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Mouse | |
| C6orf23, G6D, Ly-6C, LY6-D, Ly6G, Ly-6G, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Ly6g | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse Gr-1/Ly-6G. Weak cross-reactivity with Ly-6C is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title