missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse gp130 APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
£259.00 - £476.00
Specifications
| Antigen | gp130/CD130 |
|---|---|
| Clone | 125623 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18134081
|
R&D Systems
FAB4681A-100UG |
100 μg |
£476.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18104521
|
R&D Systems
FAB4681A-025UG |
25 μg |
£259.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
gp130/CD130 Monoclonal antibody specifically detects gp130/CD130 in Mouse samples. It is validated for Flow CytometrySpecifications
| gp130/CD130 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha | |
| Mouse myeloma cell line NS0-derived recombinant mouse gp130, Gln23-Glu617 (predicted), Accession # Q6PDI9 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. |
| 125623 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| Q6PDI9 | |
| 3572 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title