missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse GM-CSF Recombinant Protein

Recombinant Mouse Granulocyte Macrophage Colony-Stimulating Factor (GM-CSF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC2013

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10747313

  • £2975.00 / Each
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Mouse GM-CSF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of MC/9 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
14 kDa
1mg
Mouse
Carrier-Free
1 mg
E. coli
Recombinant, Ligand
GM-CSF, G-CSF, & M-CSF
Gibco™
1437
Sequential Chromatography
GM-CSF
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Wet Ice
Cytokines & Receptors
Lyophilized
Signal Transduction, Oncology, Stem Cell Research, Differentiation, Drug Development
95 %
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.