missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GM-CSF Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 12 publications
£184.00 - £1805.00
Especificaciones
| Antigen | GM-CSF |
|---|---|
| Clone | MP122E9 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.03-0.09 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18121021
![]() |
R&D Systems
MAB415-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18738829
![]() |
R&D Systems
MAB415-100 |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18764429
![]() |
R&D Systems
MAB415-500 |
500 μg |
£1805.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
GM-CSF Monoclonal specifically detects GM-CSF in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Especificaciones
| GM-CSF | |
| Western Blot 1 ug/mL, Neutralization 0.03-0.09 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P01587 | |
| 1437 | |
| Yeast derived recombinant mouse GM-CSF Accession # P01587 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse GM-CSF in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human GM-CSF. |
| MP122E9 | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim | |
| CSF2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto