missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse Glycophorin A / CD235a Protein, Fc,His Tag

Product Code. 15856357
Change view
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15856357 50 μg 50µg
15846357 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15856357 Supplier ACROBiosystems Supplier No. CDAM526X50UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycophorin-A, a member of the glycophorin-A family, is also known as CD235a and Gypa, which is heavily glycosylated on serine and threonine residues at the N-terminal extracellular domain. CD235a,a single-pass type III membrane protein,is the major intrinsic membrane sialoglycoprotein of the erythrocyte. Also,CD235a appears to be important for the function of SLC4A1 and is required for high activity of SLC4A1. Furthermore,CD235a may be involved in translocation of SLC4A1 to the plasma membrane.

This protein carries a human IgG1 Fc tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.1 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Molecular Weight (g/mol) 39.1 kDa
Quantity 50 μg
Source Mouse Glycophorin A, Fc,His Tag (CDA-M526x) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Val 108 (Accession # P14220).
Immunogen Met 1 - Val 108
Storage Requirements -20°C
Gene Alias CD235a,Glycophorin-A,Gypa
Common Name Glycophorin A
Conjugate Unconjugated
Species Mouse
Protein Tag His Tag and Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >90% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.