missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GITR Ligand/TNFSF18 Antibody, R&D Systems™

Rat Monoclonal Antibody
£156.00 - £395.00
Specifications
| Antigen | GITR Ligand/TNFSF18 |
|---|---|
| Clone | 994529 |
| Dilution | Flow Cytometry 0.25 ug/106 cells, Neutralization 1.2-12 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Neutralization, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783617
![]() |
R&D Systems
MAB21773-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742307
![]() |
R&D Systems
MAB21773-100 |
100 μg |
£395.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GITR Ligand/TNFSF18 Monoclonal antibody specifically detects GITR Ligand/TNFSF18 in Mouse samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
| GITR Ligand/TNFSF18 | |
| Flow Cytometry 0.25 ug/106 cells, Neutralization 1.2-12 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Activation-inducible TNF-related ligand, AITRL, AITRLMGC138237, GITRL, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, TNFSF18, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18, Thr47-Ser173, Accession # Q7TS55 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 994529 | |
| Flow Cytometry, Neutralization, CyTOF | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q7TS55 | |
| 8995 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title