Learn More
Mouse GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand:  R&D Systems AF1677
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18022042
  • £418.32 / 100µg
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Polyclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Immunohistochemistry.
Specifications
Specifications
GFR alpha-4/GDNF R alpha-4/Persephin receptor
Polyclonal
Unconjugated
GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor
Goat
Affinity Purified
RUO
64096
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
GFRA4
Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 Asn24-Thr237 Accession # Q99JT2
100 μg
Primary
Detects mouse GFR alpha-4/GDNF R alpha-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human GFR alpha-4 is observed.