missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GFR alpha-4/GDNF R alpha-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £452.00
Specifications
| Antigen | GFR alpha-4/GDNF R alpha-4/Persephin receptor |
|---|---|
| Clone | 171217 |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18730257
![]() |
R&D Systems
MAB1677-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18170101
|
R&D Systems
MAB1677 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFR alpha-4/GDNF R alpha-4/Persephin receptor Monoclonal specifically detects GFR alpha-4/GDNF R alpha-4/Persephin receptor in Mouse samples. It is validated for Immunohistochemistry.Specifications
| GFR alpha-4/GDNF R alpha-4/Persephin receptor | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 64096 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GFR alpha-4/GDNF R alpha-4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse GFR alpha-4/GDNF R alpha-4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse GFR alpha-2, recombinant human (rh) GFR alpha-3, recombinant rat GFR alpha-1, or rhGFR alpha-4 is observed. |
| 171217 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| GDNF family receptor alpha 4, GDNF family receptor alpha-4, GDNF R alpha-4, GDNF receptor alpha-4, GDNFR-alpha-4, GFR alpha4, GFR receptor alpha 4, GFRa-4, GFRA4, GFR-alpha-4, Persephin receptor | |
| GFRA4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title