missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-5/BMP-14 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF853
This item is not returnable.
View return policy
Description
GDF-5/BMP-14 Polyclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GDF-5/BMP-14 | |
| Polyclonal | |
| Biotin | |
| P43027 | |
| GDF5 | |
| E. coli-derived recombinant mouse GDF-5/BMP-14 Ala376-Arg495 Accession # P43027 | |
| 50 μg | |
| Primary | |
| Detects mouse GDF-5/BMP-14 in Western blots. In Western blot, approximately 20% cross-reactivity with recombinant mouse (rm) GDF-6 is observed and less than 1% cross-reactivity with rmGDF-1, rmGDF-8, and rmGDF-9 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8200 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur