Learn More
Mouse GDF-5/BMP-14 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 4 publications
Brand:  R&D Systems AF853-SP
Product Code. 18755789
  • £167.00 / 25µg
Description
GDF-5/BMP-14 Polyclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GDF-5/BMP-14
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2
Goat
Antigen Affinity-purified
RUO
8200
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
P43027
GDF5
E. coli-derived recombinant mouse GDF-5 Ala376-Arg495 Accession # P43027
25 μg
Primary
Detects mouse GDF-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse (rm) GDF-6 is observed, and less than 5% cross-reactivity with rmGDF-1, rmGDF-3, rmGDF-8, rmGDF-9, recombinant human GDF-11, and rhGDF-15 is observed.