missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-1 Propeptide Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3069-SP
This item is not returnable.
View return policy
Description
GDF-1 Polyclonal specifically detects GDF-1 in Mouse samples. It is validated for Western Blot.
Specifications
| GDF-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DORV, DTGA3, embryonic growth/differentiation factor 1, GDF1, GDF-1, growth differentiation factor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2657 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P20863 | |
| GDF1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GDF-1 Propeptide Pro25-Leu235 Accession # P20863 | |
| 25 μg | |
| Primary | |
| Detects mouse GDF-1 Propeptide in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with mature recombinant mouse (rm) GDF-1 and rmGDF-3 Propeptide is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction