missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-4 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2128
This item is not returnable.
View return policy
Description
Galectin-4 Polyclonal specifically detects Galectin-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Galectin-4 | |
| Polyclonal | |
| Unconjugated | |
| Q8K419 | |
| LGALS4 | |
| E. coli-derived recombinant mouse Galectin-4 Ala2-Ile326 Accession # Q8K419 | |
| 100 μg | |
| Primary | |
| Detects mouse Galectin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human (rh) Galectin-4, approximately 5% cross-reactivity with recombinant mouse (rm) Galectin-3, rmGalectin-7, and rhGalectin-8, and less than 1% cross-reactivity with rmGalectin-1 and rhGalectin-2 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL | |
| Antigen NY-CO-27, gal-4, GAL4, galectin 4, galectin-4, Galectin4, L-36 lactose-binding protein, L36LBP, Lactose-binding lectin 4, lectin, galactoside-binding, soluble, 4, LGALS4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3960 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction