missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Galectin-3 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
Description
The Mouse Galectin-3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Galectin-3.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Mouse |
| Conjugate | Biotin |
| Assay Range | 15.60 to 1000 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 16854 |
| Synonym | AGE-R3, Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction