missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-2 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB6667
This item is not returnable.
View return policy
Description
Galectin-2 Monoclonal specifically detects Galectin-2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Galectin-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9CQW5 | |
| LGALS2 | |
| E. coli-derived recombinant mouse Galectin-2 Ser2-Glu130 Accession # Q9CQW5 | |
| 100 μg | |
| Primary | |
| Detects mouse Galectin-2 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) Galectin-2 is observed and no cross-reactivity with rhGalectin-4, -8, -10, recombinant mouse Galectin-1, -3, -7, or -9 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 747307 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Beta-galactoside-binding lectin L-14-II, Gal-2, GAL2, galectin 2, galectin-2, Galectin2, HL14gal-2, Lactose-binding lectin 2, lectin, galactoside-binding, soluble, 2, LGALS2, MGC75071, S-Lac lectin 2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3957 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction